N-Phenyl-4-biphenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Anilinobiphenyl, 4-(Phenylamino)biphenyl, N-Phenyl-[1,1'-biphenyl]-4-amine

A164426-10G A164426-250MG A164426-25G A164426-5G A164426-1G
103805-858EA 11 USD
103805-858 76872-142 76872-144 76872-146 76872-148 77320-282
N-Phenyl-4-biphenylamine 95%
N-Phenyl-4-biphenylamine

Precaución: for Research Use Only

Formula: C₁₈H₁₅N
MW: 245.33 g/mol
Boiling Pt: 411.2 °C (760 mm Hg)
Melting Pt: 113 °C
Flash Pt: 217.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01318677
CAS Number: 32228-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR