Nortropine 95%

Supplier: Ambeed
Warning

Synonyms: (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol

A179748-100G A179748-10G A179748-1G A179748-25G A179748-5G
103807-460EA 9 USD
103807-460 103845-812 76813-516 76813-518 77083-282 77301-630 77301-632
Nortropine 95%
Nortropine

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 229.6 °C (760 mm Hg)
Melting Pt: 108…110 °C
Flash Pt: 116.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00047140
CAS Number: 538-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR