1,9-Nonanediol 98%

Supplier: Ambeed
Warning

A822535-25G A822535-1000G A822535-100G A822535-10G A822535-500G
103841-142EA 6 USD
103841-142 76845-380 77199-798 77199-800 77199-802 77199-804
1,9-Nonanediol 98%
1,9-Nonanediol

Precaución: for Research Use Only

Formula: C₉H₂₀O₂
Boiling Pt: 292.4 °C (760 mm Hg)
Melting Pt: 45…47 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00002991
CAS Number: 3937-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR