N-Methyltetrahydro-2H-pyran-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylamino)tetrahydro-2H-pyran

A124987-250MG A124987-25G A124987-1G A124987-5G A124987-10G
103800-602EA 12 USD
103800-602 76810-792 76810-794 77312-750 77312-752 77571-494
N-Methyltetrahydro-2H-pyran-4-amine 97%
N-Methyltetrahydro-2H-pyran-4-amine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
Boiling Pt: 165 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06658492
CAS Number: 220641-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR