Methyltri-n-octylammonium Hydrogen sulfate 98%

Supplier: Ambeed
Warning

Synonyms: Tri-n-octylmethylammonium Hydrogen Sulphate, N-Methyl-N,N-dioctyloctan-1-aminium hydrogensulphate

A190143-10G A190143-1G
103846-860EA 7 USD
103846-860 77111-718 77111-720
Methyltri-n-octylammonium Hydrogen sulfate 98%
Methyltri-n-octylammonium Hydrogen sulfate

Precaución: for Research Use Only

Formula: C₂₅H₅₅NO₄S
MW: 465.78 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD03095545
CAS Number: 59158-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR