(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Me-Pro-OH·H₂O, N-Methyl-L-proline Monohydrate
A575492-1G
A575492-25G
A575492-5G
103828-834EA
15
USD
103828-834
103839-982
76844-112
76844-114
76844-116
(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate 96%
(S)-1-Methylpyrrolidine-2-carboxylic acid hydrate
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₃ MW: 147.1700 g/mol Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149962 CAS Number: 199917-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...