N-Methylheptan-1-amine 97%

Supplier: Ambeed
Danger

A499692-250MG A499692-1G A499692-5G
103828-284EA 19 USD
103828-284 103839-372 76726-018 77307-616 77307-618
N-Methylheptan-1-amine 97%
N-Methylheptan-1-amine

Precaución: for Research Use Only

Formula: C₈H₁₉N
Boiling Pt: 160.8 °C (760 mm Hg)
Flash Pt: 38.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00039980
CAS Number: 36343-05-2
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR