N-Methyl-D-prolinol 95%
Supplier: Ambeed
|
Warning
|
A452537-1G
A452537-25G
A452537-5G
A452537-100G
A452537-10G
A452537-250MG
103827-800EA
12
USD
103827-800
103838-870
76842-700
76842-702
76842-704
77291-694
77291-696
77291-698
N-Methyl-D-prolinol 95%
N-Methyl-D-prolinol
Precaución: for Research Use Only
Formula:
C₆H₁₃NO MW: 115.1700 g/mol Boiling Pt: 164.8 °C (760 mm Hg) Flash Pt: 71.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368622 CAS Number: 99494-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...