N-Methylcyclopropanamine hydrochloride 98%

Supplier: Ambeed
Warning

A295281-1G A295281-5G A295281-100G
103814-050EA 5 USD
103814-050 103835-186 76837-840 76837-842 77032-404
N-Methylcyclopropanamine hydrochloride 98%
N-Methylcyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClN
MW: 107.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997746
CAS Number: 67376-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR