N-Methylbenzamide 97%

Supplier: Ambeed
Warning

A111758-10G A111758-25G A111758-1G A111758-5G A111758-100G
103797-900EA 6 USD
103797-900 103823-720 77245-728 77245-730 77310-528 77310-530 77745-827
N-Methylbenzamide 97%
N-Methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 307.5 °C (760 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: 175.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00011642
CAS Number: 613-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR