N-Methyl-N-[4-(trifluoromethyl)phenyl]amine 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-4-(trifluoromethyl)aniline

A114775-100G A114775-10G A114775-1G A114775-5G A114775-25G
103798-470EA 15 USD
103798-470 103824-312 76766-084 76766-086 77311-136 77311-138 77623-960
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine 98%
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 193 °C (760 mm Hg)
Flash Pt: 98.88 °C
Storage Temperature: Ambient
MDL Number: MFCD07369717
CAS Number: 22864-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR