N-Methyl-3-pyrrolidinol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-1-methylpyrrolidine, 1-Methyl-3-pyrrolidinol

A198212-100G A198212-10G A198212-1G A198212-25G A198212-5G
103816-998EA 7 USD
103816-998 103847-624 76999-224 76999-226 77085-640 77085-642 77085-644
N-Methyl-3-pyrrolidinol 97%
N-Methyl-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 50…52 °C (1 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003176
CAS Number: 13220-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR