2-Fluoro-N-methylaniline 98%

Supplier: Ambeed
Warning

A117102-10G A117102-25G A117102-100G
103824-704EA 5 USD
103824-704 76995-568 76995-570 77571-028
2-Fluoro-N-methylaniline 98%
2-Fluoro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈FN
Boiling Pt: 180.3 °C (760 mm Hg)
Flash Pt: 62.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00028030
CAS Number: 1978-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR