N-Methyl-1H-1,3-benzodiazol-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-1H-benzo[d]imidazol-2-amine

A144657-250MG A144657-1G
103803-434EA 188 USD
103803-434 76823-598 77316-760
N-Methyl-1H-1,3-benzodiazol-2-amine 95%
N-Methyl-1H-1,3-benzodiazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.177 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00524803
CAS Number: 17228-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR