N1-Methylbenzene-1,2-diamine dihydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-1,2-benzenediamine dihydrochloride, N-Methyl-1,2-phenylenediamine dihydrochloride

A359399-100G A359399-5G
103810-212EA 123 USD
103810-212 77278-262 77278-264
N1-Methylbenzene-1,2-diamine dihydrochloride 99%
N1-Methylbenzene-1,2-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂
MW: 195.0900 g/mol
Boiling Pt: 252.4 °C (760 mm Hg)
Melting Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00042021
CAS Number: 25148-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR