N-Methyl-1-(thiophen-2-yl)methanamine 95%
Supplier: Ambeed
|
Warning
|
A231996-1G
A231996-5G
103810-808EA
21
USD
103810-808
103850-562
77089-480
77089-482
N-Methyl-1-(thiophen-2-yl)methanamine 95%
N-Methyl-1-(thiophen-2-yl)methanamine
Precaución: for Research Use Only
Formula:
C₆H₉NS Boiling Pt: 170.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD04625435 CAS Number: 58255-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...