3-(Methylaminomethyl)pyridine 98%

Supplier: Ambeed
Warning

A102178-10G A102178-1G
103796-326EA 33 USD
103796-326 103822-020 77093-556 77308-894
3-(Methylaminomethyl)pyridine 98%
3-(Methylaminomethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
Boiling Pt: 198.2 °C (760 mm Hg)
Melting Pt: 21 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023610
CAS Number: 20173-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR