N-Methyl-1-(p-tolyl)methanamine 97%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-4-methylbenzylamine

A161959-10G A161959-25G A161959-1G A161959-5G
103805-556EA 6 USD
103805-556 103843-800 76720-150 77300-566 77319-818 77319-820
N-Methyl-1-(p-tolyl)methanamine 97%
N-Methyl-1-(p-tolyl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.2100 g/mol
Flash Pt: 76.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115412
CAS Number: 699-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR