N-Methyl-2-methylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-1-(o-tolyl)methanamine

A652136-1G A652136-5G
103840-402EA 88 USD
103840-402 77308-064 77308-066
N-Methyl-2-methylbenzylamine 97%
N-Methyl-2-methylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 196.5 °C (760 mm Hg)
Flash Pt: 78.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04625428
CAS Number: 874-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR