N-Methyl-1-naphthylmethylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Methylaminomethyl)naphthalene

A127846-100G A127846-10G A127846-500G A127846-25G A127846-5G
103808-362EA 157 USD
103808-362 76718-826 76718-828 76718-830 77298-236 77313-300
N-Methyl-1-naphthylmethylamine 98%
N-Methyl-1-naphthylmethylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Flash Pt: 146.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00144934
CAS Number: 14489-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR