N-Methoxy-N-methylpropionamide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-Methoxy-N-methylpropanamide
A100208-250MG
A100208-25G
A100208-1G
A100208-5G
A100208-10G
103795-296EA
13
USD
103795-296
103821-008
76717-470
76717-472
77296-594
77296-596
77570-006
N-Methoxy-N-methylpropionamide 97%
N-Methoxy-N-methylpropionamide
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09833541 CAS Number: 104863-65-2 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...