N-Methoxy-N,2-dimethylpropanamide 98%

Supplier: Ambeed
Warning

Synonyms: N-Methoxy-N-methylisobutyramide

A458240-100G A458240-10G A458240-1G A458240-25G A458240-5G
103827-888EA 9 USD
103827-888 103838-964 76829-786 76829-788 76842-832 76842-834 76842-836
N-Methoxy-N,2-dimethylpropanamide 98%
N-Methoxy-N,2-dimethylpropanamide

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.173 g/mol
Boiling Pt: 137.7±23.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11847606
CAS Number: 113778-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR