2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione 98%

Supplier: Ambeed
Warning

Synonyms: Nitisinone, 2-(2-Nitro-4-(trifluoromethyl)benzoyl)cyclohexane-1,3-dione

A384235-25G A384235-50MG A384235-5G A384235-10G A384235-10MG A384235-1MG A384235-250MG
103799-636EA 14 USD
103799-636 103826-756 103837-792 76841-136 76841-138 76841-140 77240-438 77240-440 77240-442 77240-444
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione 98%
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃NO₅
MW: 329.231 g/mol
Boiling Pt: 486.2 °C (760 mm Hg)
Melting Pt: 129…131 °C
Flash Pt: 247.9 °C
Storage Temperature: Freezer
MDL Number: MFCD01752192
CAS Number: 104206-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR