N-Isopropyl chloroacetamide 97%

Supplier: Ambeed
Warning

A327967-1G A327967-5G
103825-484EA 27 USD
103825-484 103836-436 76839-404 76839-406
N-Isopropyl chloroacetamide 97%
N-Isopropyl chloroacetamide

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
Boiling Pt: 250.6 °C (760 mm Hg)
Melting Pt: 58…60 °C (760 mm Hg)
Flash Pt: 105.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068034
CAS Number: 2895-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR