N2-Isobutyrylguanosine 95%

Supplier: Ambeed
Warning

Synonyms: N-Isobutyrylguanosine, N-(9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide

A166360-100G A166360-10G A166360-1G A166360-25G A166360-5G
103806-058EA 6 USD
103806-058 103844-320 76720-320 76720-322 77320-566 77320-568 77320-570
N2-Isobutyrylguanosine 95%
N2-Isobutyrylguanosine

Precaución: for Research Use Only

Formula: C₁₄H₁₉N₅O₆
MW: 353.34 g/mol
Melting Pt: 148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274102
CAS Number: 64350-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR