2-(3-Trifluoromethylanilino)nicotinic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-[(3-Trifluoromethylphenyl)amino]nicotinic Acid, 2-((3-(Trifluoromethyl)phenyl)amino)nicotinic acid, Niflumic Acid

A207771-100MG A207771-1G A207771-5G
103817-824EA 16 USD
103817-824 103848-508 76975-410 77086-790 77086-792
2-(3-Trifluoromethylanilino)nicotinic acid 98%
2-(3-Trifluoromethylanilino)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₉F₃N₂O₂
MW: 282.22 g/mol
Boiling Pt: 378 °C (760 mm Hg)
Melting Pt: 203…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00010569
CAS Number: 4394-00-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR