Nickel(II) acetylacetonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate
A167207-25G
A167207-100G
A167207-10G
A167207-500G
103808-982EA
104
USD
103808-982
103844-410
77081-410
77110-882
77110-884
77110-886
Nickel(II) acetylacetonate 97%
Nickel(II) acetylacetonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄NiO₄ MW: 256.91 g/mol Boiling Pt: 220 Melting Pt: 208 Flash Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000024 CAS Number: 3264-82-2 Merck Index: 12,06584 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...