Nevirapine 98%

Supplier: Ambeed
Warning

Synonyms: 11-Cyclopropyl-4-methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one

A326622-25G A326622-5G A326622-250MG A326622-1G A326622-100G
103825-442EA 8 USD
103825-442 103836-390 76839-348 76839-350 77114-820 77306-350 77685-936
Nevirapine 98%
Nevirapine

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₄O
MW: 266.30 g/mol
Boiling Pt: 415.4 °C (760 mm Hg)
Melting Pt: 247 °C
Flash Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00866928
CAS Number: 129618-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR