N-Ethyl-N-methylcarbamoyl chloride 98%
Supplier: Ambeed
|
Danger
|
A435817-5G
A435817-100G
A435817-10G
A435817-25G
103838-668EA
6
USD
103838-668
76842-396
76979-340
76979-342
76979-344
77307-290
N-Ethyl-N-methylcarbamoyl chloride 98%
N-Ethyl-N-methylcarbamoyl chloride
Precaución: For research use only
Formula:
C₄H₈ClNO Boiling Pt: 165.1 °C (760 mm Hg) Flash Pt: 53.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08061404 CAS Number: 42252-34-6 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...