N-Cyclohexylaniline 97%

Supplier: Ambeed
Warning

A176840-100G A176840-10G A176840-1G A176840-25G A176840-5G
103807-174EA 6 USD
103807-174 103845-502 76720-708 76720-710 77082-860 77082-862 77082-864
N-Cyclohexylaniline 97%
N-Cyclohexylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₇N
Boiling Pt: 292.9 °C (760 mm Hg)
Melting Pt: 14…15 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00014284
CAS Number: 1821-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR