N-Cyano-N-phenyl-p-toluenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: N-Cyano-4-methyl-N-phenylbenzenesulphonamide

A111494-100MG A111494-250MG A111494-1G A111494-5G A111494-25G
103797-848EA 16 USD
103797-848 103823-668 77160-764 77160-766 77310-470 77310-472 77683-020
N-Cyano-N-phenyl-p-toluenesulfonamide 95%
N-Cyano-N-phenyl-p-toluenesulfonamide

Precaución: For research use only.

Formula: C₁₄H₁₂N₂O₂S
MW: 272.322 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00159358
CAS Number: 55305-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR