N-Carbobenzoxy-L-threonine 96%

Supplier: Ambeed
Warning

Synonyms: Z-Thr-OH, (2S,3R)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxybutanoic acid, N-Cbz-L-threonine

A355730-100G A355730-10G
103837-156EA 8 USD
103837-156 77278-220 77278-222
N-Carbobenzoxy-L-threonine 96%
N-Carbobenzoxy-L-threonine

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₅
MW: 253.25 g/mol
Boiling Pt: 483.7 °C (760 mm Hg)
Melting Pt: 101…103 °C
Storage Temperature: Freezer
MDL Number: MFCD00065948
CAS Number: 19728-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR