N-Cbz-4-piperidinecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyloxycarbonyl-4-piperidinecarboxylic acid, 1-Benzyloxycarbonylisonipecotic acid, 1-Carbobenzoxy-4-piperidinecarboxylic acid, 1-Cbz-isonipecotic acid, 1-Carbobenzoxyisonipecotic acid, 1-Cbz-4-piperidinecarboxylic acid, N-Cbz-4-Piperidinecarboxylic acid

A204387-100G A204387-10G A204387-1G A204387-25G A204387-5G A204387-500G
103817-550EA 5 USD
103817-550 103848-210 76785-446 76785-448 77086-378 77086-380 77086-382 77667-708
N-Cbz-4-piperidinecarboxylic acid 95%
N-Cbz-4-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
Boiling Pt: 443.9 °C (760 mm Hg)
Melting Pt: 78 °C
Flash Pt: 222.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01568759
CAS Number: 10314-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR