N-Cbz-2-piperidinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyloxycarbonylpipecolic acid, 1-Carbobenzoxypipecolic acid, 1-Carbobenzoxy-2-piperidinecarboxylic acid, N-Cbz-2-Piperidinecarboxylic acid, Z-DL-Pip-OH, 1-Benzyloxycarbonyl-2-piperidinecarboxylic acid, 1-Cbz-pipecolic acid, 1-Cbz-2-piperidinecarboxylic acid

A205730-100G A205730-500G A205730-5G A205730-25G
103848-322EA 7 USD
103848-322 76785-482 76785-484 77086-550 77302-962
N-Cbz-2-piperidinecarboxylic acid 98%
N-Cbz-2-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.2900 g/mol
Melting Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863568
CAS Number: 28697-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR