N-Boc-pyrrole 95%

Supplier: Ambeed
Warning

A657885-100G A657885-10G A657885-25G A657885-5G A657885-500G
103840-426EA 12 USD
103840-426 76727-410 76727-412 76844-614 76844-616 77880-383
N-Boc-pyrrole 95%
N-Boc-pyrrole

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
Boiling Pt: 223.2 °C (760 mm Hg)
Flash Pt: 88.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00209559
CAS Number: 5176-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR