N-(tert-Butoxycarbonyl)propargylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-propargylamine, 2-Propynylcarbamic Acid tert-Butyl Ester, tert-Butyl 2-Propynylcarbamate

A171481-100G A171481-10G A171481-500G A171481-25G A171481-5G A171481-1G
103806-638EA 7 USD
103806-638 103844-912 77011-588 77011-590 77011-592 77082-076 77082-078 77301-132
N-(tert-Butoxycarbonyl)propargylamine 98%
N-(tert-Butoxycarbonyl)propargylamine

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
Melting Pt: 40…44 °C
Flash Pt: 93.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367245
CAS Number: 92136-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR