N-Boc-morpholine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Morpholine-2-carboxylic acid, N-BOC protected

A102703-100G A102703-10G A102703-250MG A102703-25G A102703-500G A102703-1G A102703-5G
103796-414EA 6 USD
103796-414 103822-112 77282-960 77282-962 77282-964 77282-966 77282-968 77308-980 77308-982
N-Boc-morpholine-2-carboxylic acid 97%
N-Boc-morpholine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.2500 g/mol
Boiling Pt: 369.5 °C (760 mm Hg)
Flash Pt: 177.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321006
CAS Number: 189321-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR