N-Boc-L-prolinol 98%

Supplier: Ambeed
Warning

A137477-100G A137477-10G A137477-25G A137477-5G A137477-500G
76811-594EA 55 USD
76811-594 76811-596 77315-422 77315-424 77670-970
N-Boc-L-prolinol 98%
N-Boc-L-prolinol

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
Boiling Pt: 289.5 °C (760 mm Hg)
Melting Pt: 62…64 °C
Flash Pt: 128.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066232
CAS Number: 69610-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR