N-Boc-D-prolinal 97%

Supplier: Ambeed
Warning

Synonyms: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate

A126248-100G A126248-10G A126248-1G A126248-5G
103800-780EA 14 USD
103800-780 76718-730 76718-732 77312-976 77312-978
N-Boc-D-prolinal 97%
N-Boc-D-prolinal

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
Boiling Pt: 276.3 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD01321389
CAS Number: 73365-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR