N-Boc-DL-valinol 97%
Supplier: Ambeed
|
Warning
|
A408294-100MG
A408294-1G
103827-182EA
35
USD
103827-182
103838-228
76725-152
76841-738
N-Boc-DL-valinol 97%
N-Boc-DL-valinol
Precaución: for Research Use Only
Formula:
C₁₀H₂₁NO₃ MW: 203.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01456557 CAS Number: 169556-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...