cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95%

Supplier: Ambeed
Warning

Synonyms: Boc-cis-D-Hyp-OH, (2R,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, (2R,4R)-N-Boc-4-hydroxy-2-pyrrolidinecarboxylic acid

A159510-10G A159510-1G A159510-25G A159510-5G A159510-100G A159510-500G
103805-258EA 6 USD
103805-258 103843-478 76997-736 77300-404 77319-364 77319-366 77592-940 77881-248
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95%
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
Melting Pt: 146…151 °C
Flash Pt: 190.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094407
CAS Number: 135042-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR