N-Boc-5-hydroxyindole 95%

Supplier: Ambeed
Warning

A434477-100MG A434477-250MG A434477-1G A434477-10G
103827-574EA 69 USD
103827-574 103838-640 76979-322 76979-324 77307-276 77594-350
N-Boc-5-hydroxyindole 95%
N-Boc-5-hydroxyindole

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
Boiling Pt: 374.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08272246
CAS Number: 434958-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR