N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(Fmoc-amino)-piperidine-4-carboxylic acid, N-Boc-4-(Fmoc-amino)piperidine-4-carboxylic acid

A266792-10G A266792-250MG A266792-1G A266792-5G
103812-814EA 27 USD
103812-814 103852-716 77001-038 77001-040 77092-420 77092-422
N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid 97%
N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₆H₃₀N₂O₆
Boiling Pt: 657.8 °C (760 mm Hg)
Melting Pt: 160…164 °C
Flash Pt: 351.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318741
CAS Number: 183673-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR