tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: N-BOC-3-pyrrolidinone

A291375-25G A291375-5G A291375-100G A291375-10G A291375-1G
103813-912EA 6 USD
103813-912 103835-052 76837-644 76837-646 77032-304 77032-306 77305-610
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Boiling Pt: 270.9 °C (760 mm Hg)
Melting Pt: 34…38 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631194
CAS Number: 101385-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR