N-Boc-3-methylenepiperidine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-Methylenepiperidine

A222006-1G A222006-5G
103818-962EA 15 USD
103818-962 103849-838 77088-436 77088-438
N-Boc-3-methylenepiperidine 98%
N-Boc-3-methylenepiperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MW: 197.2741 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706141
CAS Number: 276872-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR