N-Boc-3-cyanopyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-Cyanopyrrolidine

A144438-10G A144438-25G A144438-5G A144438-1G
103803-406EA 26 USD
103803-406 103841-482 76689-116 77299-382 77299-384 77316-726
N-Boc-3-cyanopyrrolidine 97%
N-Boc-3-cyanopyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.2462 g/mol
Boiling Pt: 307.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03001709
CAS Number: 476493-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR