N-Boc-3,5-difluoro-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(3,5-DiF)-OH

A193260-250MG A193260-1G A193260-5G
103816-518EA 115 USD
103816-518 103847-132 76785-000 77085-028 77085-030
N-Boc-3,5-difluoro-L-phenylalanine 98%
N-Boc-3,5-difluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₇F₂NO₄
MW: 301.29 g/mol
Boiling Pt: 417.6 °C (760 mm Hg)
Melting Pt: 115 °C
Flash Pt: 206.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00797555
CAS Number: 205445-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR