4-(tert-Butoxycarbonyl)thiomorpholine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: N-BOC-thiomorpholine-2-carboxylic acid

A288366-250MG A288366-100MG
103813-790EA 135 USD
103813-790 77593-768 77879-564
4-(tert-Butoxycarbonyl)thiomorpholine-2-carboxylic acid ≥95%
4-(tert-Butoxycarbonyl)thiomorpholine-2-carboxylic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₀H₁₇NO₄S
Boiling Pt: 398.6 °C (760 mm Hg)
Flash Pt: 194.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426523
CAS Number: 134676-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR