N-Boc-2-pyrroleboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrole-2-boronic acid, N-BOC protected, N-Boc-2-Pyrroleboronic acid

A219229-1G A219229-25G A219229-5G A219229-10G A219229-100G
103818-774EA 7 USD
103818-774 103849-646 77088-194 77088-196 77088-198 77249-340 77593-426
N-Boc-2-pyrroleboronic acid 98%
N-Boc-2-pyrroleboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₄
MW: 211.02 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Freezer
MDL Number: MFCD01318939
CAS Number: 135884-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR