N-Boc-2-chloroethylamine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloroethylamine, N-BOC protected

A164103-10G A164103-1G A164103-25G A164103-5G
103805-816EA 7 USD
103805-816 103844-080 76783-996 77320-220 77320-222 77320-224
N-Boc-2-chloroethylamine 98%
N-Boc-2-chloroethylamine

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.6446 g/mol
Flash Pt: 103.1 °C
Storage Temperature: Freezer
MDL Number: MFCD04004074
CAS Number: 71999-74-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR